Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685123.3 | XP_005685180.1 | 2 - 3 | 383 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |