Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059750.1 | XP_017915239.1 | 7 - 8 | 801 | 67 | |
GenBank mRNA | KA309286.1 | - | 7 - 8 | 789 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |