Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693302.3 | XP_005693359.1 | 2 - 3 | 580 | 95 | |
GenBank mRNA | KA323109.1 | - | 2 - 3 | 572 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |