Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693407.3 | XP_005693464.2 | 5 - 6 | 910 | 80 | |
GenBank mRNA | JO428995.2 | - | 1 - 2 | 1376 | 80 | |
JT675986.1 | - | 6 - 7 | 889 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |