Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695881.3 | XP_005695938.2 | 4 - 5 | 816 | 115 | |
XM_018066936.1 | XP_017922425.1 | 4 - 5 | 2046 | 115 | ||
GenBank mRNA | JP351349.2 | - | 3 - 4 | 969 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Probe spans exons |