Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696300.3 | XP_005696357.1 | 2 - 3 | 476 | 89 | |
GenBank mRNA | JO639225.2 | - | 2 - 3 | 461 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |