Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054851.1 | XP_017910340.1 | 2 - 3 | 219 | 118 | |
GenBank mRNA | KA344434.1 | - | 2 - 3 | 174 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |