Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063699.1 | XP_017919188.1 | 14 - 15 | 2506 | 67 | |
XM_018063700.1 | XP_017919189.1 | 14 - 15 | 2534 | 67 | ||
XM_018063701.1 | XP_017919190.1 | 14 - 15 | 2458 | 67 | ||
XM_018063702.1 | XP_017919191.1 | 14 - 15 | 2437 | 67 | ||
GenBank mRNA | JO577302.1 | - | 2 - 3 | 683 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |