Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038779.1 | XP_017894268.1 | 6 - 7 | 868 | 74 | |
XM_018038780.1 | XP_017894269.1 | 6 - 7 | 770 | 74 | ||
XM_018038781.1 | XP_017894270.1 | 6 - 7 | 868 | 74 | ||
XM_018038782.1 | XP_017894271.1 | 6 - 7 | 770 | 74 | ||
GenBank mRNA | JR126207.1 | - | 3851 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |