Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEPCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013974943.2 | XP_013830397.2 | 2 - 3 | 2490 | 65 | |
GenBank mRNA | JI861724.1 | - | 2 - 3 | 481 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |