Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XKR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041815.1 | XP_017897304.1 | 2 - 3 | 653 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |