Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042200.1 | XP_017897689.1 | 2 - 3 | 460 | 95 | |
GenBank mRNA | JO344206.2 | - | 2 - 3 | 456 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |