Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674692.3 | XP_005674749.2 | 1 - 2 | 881 | 71 | |
GenBank mRNA | JO353076.2 | - | 6 - 7 | 678 | 71 | |
JO375642.2 | - | 3 - 4 | 303 | 71 | ||
JO415835.2 | - | 6 - 7 | 681 | 71 | ||
JO437008.2 | - | 1 - 2 | 377 | 71 | ||
JO504230.2 | - | 1 - 2 | 289 | 71 | ||
JO594966.2 | - | 1 - 2 | 369 | 71 | ||
JO631207.2 | - | 1 - 2 | 331 | 71 | ||
JT659292.1 | - | 1 - 2 | 36 | 71 | ||
JT705606.1 | - | 1 - 2 | 149 | 71 | ||
JT717661.1 | - | 1 - 2 | 175 | 71 | ||
JT738669.1 | - | 1 - 2 | 137 | 71 | ||
KA321950.1 | - | 1 - 2 | 464 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |