Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963501.2 | XP_013818955.1 | 3 - 4 | 612 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |