Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066346.1 | XP_017921835.1 | 13 - 14 | 2561 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |