Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681228.2 | XP_005681285.2 | 3 - 4 | 525 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |