Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682107.3 | XP_005682164.2 | 4 - 5 | 1646 | 57 | |
XM_005682108.3 | XP_005682165.2 | 2 - 3 | 1228 | 57 | ||
GenBank mRNA | JO611415.2 | - | 1 - 2 | 434 | 57 | |
JT634382.1 | - | 1 - 2 | 435 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |