Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683438.2 | XP_005683495.1 | 9 - 10 | 1224 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |