Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686811.3 | XP_005686868.1 | 2 - 3 | 248 | 93 | |
XM_018055436.1 | XP_017910925.1 | 2 - 3 | 256 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |