Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692196.3 | XP_005692253.1 | 3 - 4 | 436 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |