Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692819.3 | XP_005692876.2 | 3 - 4 | 572 | 75 | |
XM_013971511.2 | XP_013826965.1 | 3 - 4 | 537 | 75 | ||
XM_013971512.2 | XP_013826966.1 | 3 - 4 | 256 | 75 | ||
GenBank mRNA | JO581400.1 | - | 3 - 4 | 658 | 75 | |
JO581401.1 | - | 3 - 4 | 655 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |