Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055267.1 | XP_017910756.1 | 3 - 4 | 2148 | 72 | |
XM_018055268.1 | XP_017910757.1 | 3 - 4 | 2061 | 72 | ||
XM_018055269.1 | XP_017910758.1 | 3 - 4 | 2148 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |