Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC85A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055267.1 | XP_017910756.1 | 5 - 6 | 2406 | 76 | |
XM_018055268.1 | XP_017910757.1 | 5 - 6 | 2319 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |