Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677897.3 | XP_005677954.1 | 6 - 7 | 792 | 72 | |
GenBank mRNA | JO437631.2 | - | 6 - 7 | 828 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |