Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687208.3 | XP_005687265.1 | 2 - 3 | 280 | 67 | |
GenBank mRNA | JO584870.2 | - | 2 - 3 | 254 | 67 | |
KA316541.1 | - | 2 - 3 | 272 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |