Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687208.3 | XP_005687265.1 | 4 - 5 | 525 | 96 | |
XM_005687209.3 | XP_005687266.1 | 4 - 5 | 417 | 96 | ||
GenBank mRNA | JO584870.2 | - | 4 - 5 | 499 | 96 | |
KA316541.1 | - | 4 - 5 | 517 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |