Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968059.2 | XP_013823513.2 | 2 - 3 | 460 | 84 | |
XM_018056141.1 | XP_017911630.1 | 4 - 5 | 567 | 84 | ||
XM_018056142.1 | XP_017911631.1 | 4 - 5 | 536 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |