Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040922.1 | XP_017896411.1 | 3 - 4 | 1236 | 103 | |
GenBank mRNA | JO632797.2 | - | 3 - 4 | 1271 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Capra hircus Probe spans exons |