Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPSE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698313.2 | XP_005698370.2 | 3 - 4 | 738 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |