Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963641.2 | XP_013819095.2 | 20 - 21 | 2051 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |