Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039294.1 | XP_017894783.1 | 7 - 8 | 1558 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |