Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FECH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039721.1 | XP_017895210.1 | 8 - 9 | 1000 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |