Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013976239.2 | XP_013831693.2 | 4 - 5 | 893 | 56 | |
GenBank mRNA | JO583980.1 | - | 2 - 3 | 525 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |