Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013976267.2 | XP_013831721.2 | 1 - 2 | 943 | 75 | |
XM_018044214.1 | XP_017899703.1 | 1 - 2 | 944 | 75 | ||
GenBank mRNA | JO620511.2 | - | 1 - 2 | 70 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |