Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677515.3 | XP_005677572.2 | 7 - 8 | 1368 | 84 | |
XM_018046127.1 | XP_017901616.1 | 7 - 8 | 1458 | 84 | ||
XM_018046128.1 | XP_017901617.1 | 7 - 8 | 1470 | 84 | ||
XM_018046129.1 | XP_017901618.1 | 7 - 8 | 1413 | 84 | ||
XM_018046131.1 | XP_017901620.1 | 6 - 7 | 1221 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |