Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680857.3 | XP_005680914.2 | 2 - 3 | 211 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |