Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686226.2 | XP_005686283.2 | 1 - 2 | 83 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |