Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686803.3 | XP_005686860.1 | 3 - 4 | 2938 | 88 | |
XM_018055427.1 | XP_017910916.1 | 4 - 5 | 2103 | 88 | ||
XM_018055428.1 | XP_017910917.1 | 4 - 5 | 2103 | 88 | ||
GenBank mRNA | JO631208.2 | - | 4 - 5 | 2121 | 88 | |
JO631212.2 | - | 4 - 5 | 2121 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |