Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686803.3 | XP_005686860.1 | 5 - 6 | 3167 | 69 | |
XM_018055427.1 | XP_017910916.1 | 6 - 7 | 2332 | 69 | ||
XM_018055428.1 | XP_017910917.1 | 6 - 7 | 2332 | 69 | ||
GenBank mRNA | JO631208.2 | - | 6 - 7 | 2350 | 69 | |
JO631212.2 | - | 6 - 7 | 2350 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |