Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689227.3 | XP_005689284.1 | 3 - 4 | 547 | 68 | |
XM_013969275.2 | XP_013824729.1 | 2 - 3 | 2846 | 68 | ||
XM_018058240.1 | XP_017913729.1 | 3 - 4 | 2211 | 68 | ||
XM_018058241.1 | XP_017913730.1 | 3 - 4 | 1310 | 68 | ||
XM_018058242.1 | XP_017913731.1 | 2 - 3 | 436 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |