Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018067017.1 | XP_017922506.1 | 2 - 3 | 248 | 77 | |
GenBank mRNA | JO610784.2 | - | 2 - 3 | 120 | 77 | |
JT680277.1 | - | 2 - 3 | 94 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |