Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047287.1 | XP_017902776.1 | 2 - 3 | 1554 | 69 | |
GenBank mRNA | JT732032.1 | - | 1 - 2 | 49 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Capra hircus Probe spans exons |