Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967537.2 | XP_013822991.1 | 9 - 10 | 927 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |