Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056567.1 | XP_017912056.1 | 10 - 11 | 1306 | 89 | |
GenBank mRNA | JT681352.1 | - | 7 - 8 | 3440 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |