Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058083.1 | XP_017913572.1 | 4 - 5 | 2078 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |