Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061204.1 | XP_017916693.1 | 5 - 6 | 696 | 77 | |
GenBank mRNA | JO405320.2 | - | 2 - 3 | 311 | 77 | |
KA351649.1 | - | 4 - 5 | 1044 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |