Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681101.3 | XP_005681158.1 | 9 - 10 | 1456 | 66 | |
XM_013964365.2 | XP_013819819.1 | 10 - 11 | 1604 | 66 | ||
GenBank mRNA | JO604556.2 | - | 1 - 2 | 393 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |