Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966815.2 | XP_013822269.1 | 4 - 5 | 1470 | 85 | |
XM_018054655.1 | XP_017910144.1 | 4 - 5 | 1490 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |