Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C13H20orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688312.1 | XP_005688369.1 | 1 - 2 | 118 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |