Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692684.3 | XP_005692741.2 | 8 - 9 | 1713 | 78 | |
XM_018062727.1 | XP_017918216.1 | 7 - 8 | 1526 | 78 | ||
GenBank mRNA | JO633228.2 | - | 8 - 9 | 1654 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |